Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lsm5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007645382.1 | XP_007643572.1 | 2 - 3 | 217 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Probe spans exons |