Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hoxd13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003498319.1 | XP_003498367.1 | 1 - 2 | 366 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Probe spans exons |