Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrpl32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007646920.1 | XP_007645110.1 | 1 - 2 | 200 | 93 | |
GenBank mRNA | JI886647.1 | - | 1 - 2 | 175 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Probe spans exons |