Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003508846.2 | XP_003508894.1 | 2 - 3 | 618 | 77 | |
XM_007649459.1 | XP_007647649.1 | 2 - 3 | 618 | 77 | ||
XM_007649460.1 | XP_007647650.1 | 3 - 4 | 719 | 77 |