Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003508846.2 | XP_003508894.1 | 4 - 5 | 956 | 69 | |
XM_007649459.1 | XP_007647649.1 | 4 - 5 | 956 | 69 | ||
XM_007649460.1 | XP_007647650.1 | 5 - 6 | 1057 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Probe spans exons |