Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003499480.2 | XP_003499528.1 | 6 - 7 | 1076 | 135 | |
XM_007640219.1 | XP_007638409.1 | 6 - 7 | 1110 | 135 | ||
XM_007640220.1 | XP_007638410.1 | 6 - 7 | 1110 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |