Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gimap8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003501330.1 | XP_003501378.1 | 4 - 5 | 1415 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Probe spans exons |