Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nsun3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007609741.1 | XP_007607931.1 | 2 - 3 | 162 | 116 | |
GenBank mRNA | JI864047.1 | - | 2 - 3 | 168 | 116 | |
JP042244.1 | - | 2 - 3 | 168 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |