Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rgs17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003495881.2 | XP_003495929.2 | 2 - 3 | 277 | 81 | |
XM_007644918.1 | XP_007643108.1 | 2 - 3 | 140 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Probe spans exons |