Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tti2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003497839.2 | XP_003497887.1 | 1 - 2 | 1030 | 102 | |
GenBank mRNA | JI890284.1 | - | 5 - 6 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |