Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sln | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003508785.2 | XP_003508833.1 | 1 | 317 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Both primers and probe map within a single exon |