Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rgmb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007645799.1 | XP_007643989.1 | 3 - 4 | 680 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Probe spans exons |