Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Insl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007644955.1 | XP_007643145.1 | 2 - 3 | 198 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Probe spans exons |