Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bad | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003511281.2 | XP_003511329.1 | 3 - 4 | 790 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Apoptosis Probe spans exons |