Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dcakd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003504511.2 | XP_003504559.1 | 2 - 3 | 276 | 63 | |
XM_007645059.1 | XP_007643249.1 | 2 - 3 | 262 | 63 | ||
XM_007645060.1 | XP_007643250.1 | 2 - 3 | 360 | 63 | ||
XM_007645061.1 | XP_007643251.1 | 2 - 3 | 485 | 63 | ||
GenBank mRNA | JI862990.1 | - | 1 - 2 | 215 | 63 | |
JP054718.1 | - | 1 - 2 | 214 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Probe spans exons |