Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Znf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003510416.2 | XP_003510464.1 | 4 - 5 | 576 | 92 | |
XM_007651034.1 | XP_007649224.1 | 4 - 5 | 826 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Probe spans exons |