Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnni2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007650897.1 | XP_007649087.1 | 6 - 7 | 556 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Probe spans exons |