Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gzmm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003502598.2 | XP_003502646.1 | 3 - 4 | 1505 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Protease 3' Most Probe spans exons |