Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Txlng | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003509272.2 | XP_003509320.1 | 2 - 3 | 450 | 73 | |
GenBank mRNA | JI891021.1 | - | 2 - 3 | 421 | 73 | |
JP038795.1 | - | 2 - 3 | 421 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Probe spans exons |