Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100754798 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007643250.1 | XP_007641440.1 | 5 - 6 | 787 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Probe spans exons |