Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rxfp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007643545.1 | XP_007641735.1 | 1 | 1826 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Both primers and probe map within a single exon |