Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nlrx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003511554.2 | XP_003511602.1 | 4 - 5 | 504 | 55 | |
XM_007652112.1 | XP_007650302.1 | 4 - 5 | 785 | 55 | ||
XM_007652113.1 | XP_007650303.1 | 4 - 5 | 602 | 55 | ||
XM_007652114.1 | XP_007650304.1 | 5 - 6 | 645 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Probe spans exons |