Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cartpt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003503682.2 | XP_003503730.1 | 1 - 2 | 162 | 53 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Probe spans exons |