Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cideb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003502087.2 | XP_003502135.2 | 6 - 7 | 833 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Apoptosis 3' Most Detect Genomic DNA Probe spans exons |