Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sssca1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003513905.2 | XP_003513953.1 | 3 - 4 | 322 | 56 | |
GenBank mRNA | JI883492.1 | - | 1 - 2 | 86 | 56 | |
JP057908.1 | - | 3 - 4 | 337 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Probe spans exons |