Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dohh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007646962.1 | XP_007645152.1 | 3 - 4 | 684 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Probe spans exons |