Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003501811.1 | XP_003501859.1 | 1 - 2 | 419 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Apoptosis 3' Most Recommended Assay Probe spans exons |