Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scarf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003514778.2 | XP_003514826.1 | 5 - 6 | 1349 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Probe spans exons |