Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pfn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003497693.2 | XP_003497741.1 | 3 - 4 | 399 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Probe spans exons |