Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gspt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003505683.2 | XP_003505731.1 | 1 | 724 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Both primers and probe map within a single exon |