Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fdx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003502757.2 | XP_003502805.1 | 3 - 4 | 594 | 70 | |
GenBank mRNA | JI885032.1 | - | 3 - 4 | 291 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Probe spans exons |