Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sgpp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003512336.2 | XP_003512384.2 | 2 - 3 | 227 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Apoptosis Probe spans exons |