Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gltp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007644147.1 | XP_007642337.1 | 5 - 6 | 610 | 72 | |
GenBank mRNA | JI886711.1 | - | 2 - 3 | 299 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Probe spans exons |