Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dok1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003511127.2 | XP_003511175.1 | 3 - 4 | 914 | 70 | |
XM_007651678.1 | XP_007649868.1 | 2 - 3 | 620 | 70 | ||
GenBank mRNA | JI885589.1 | - | 2 - 3 | 157 | 70 | |
JP045354.1 | - | 2 - 3 | 157 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Probe spans exons |