Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zrsr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007652072.1 | XP_007650262.1 | 9 - 10 | 1159 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |