Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003502888.2 | XP_003502936.2 | 2 - 3 | 430 | 81 | |
GenBank mRNA | JP062988.1 | - | 2 - 3 | 203 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Probe spans exons |