Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lyrm5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003498810.2 | XP_003498858.1 | 1 - 2 | 967 | 85 | |
XM_007633455.1 | XP_007631645.1 | 2 - 3 | 169 | 85 | ||
XM_007633459.1 | XP_007631649.1 | 2 - 3 | 162 | 85 | ||
GenBank mRNA | JP037679.1 | - | 2 - 3 | 169 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Probe spans exons |