Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gfi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007642991.1 | XP_007641181.1 | 5 - 6 | 694 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Probe spans exons |