Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc45a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003501206.2 | XP_003501254.2 | 2 - 3 | 952 | 51 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Probe spans exons |