Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klrg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003509228.1 | XP_003509276.1 | 2 - 3 | 188 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Probe spans exons |