Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp91 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007654606.1 | XP_007652796.1 | 9 - 10 | 848 | 85 | |
GenBank mRNA | JP055359.1 | - | 9 - 10 | 839 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Probe spans exons |