Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anks4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003513208.1 | XP_003513256.1 | 1 - 2 | 169 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Probe spans exons |