Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Igfbp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003515025.2 | XP_003515073.1 | 2 - 3 | 479 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Probe spans exons |