Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fibcd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003513748.1 | XP_003513796.1 | 6 - 7 | 1130 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Probe spans exons |