Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pglyrp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003504725.2 | XP_003504773.2 | 5 - 6 | 1656 | 63 | |
XM_007645225.1 | XP_007643415.1 | 6 - 7 | 1968 | 63 | ||
XM_007645226.1 | XP_007643416.1 | 6 - 7 | 1854 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Probe spans exons |