Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lacc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003500302.2 | XP_003500350.1 | 5 - 6 | 1335 | 71 | |
GenBank mRNA | JI863040.1 | - | 5 - 6 | 1292 | 71 | |
JP057077.1 | - | 5 - 6 | 1292 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Probe spans exons |