Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp1r3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003496522.1 | XP_003496570.1 | 2 - 3 | 850 | 100 | |
XM_007650933.1 | XP_007649123.1 | 2 - 3 | 847 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Probe spans exons |