Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glce | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003498933.2 | XP_003498981.1 | 4 - 5 | 962 | 78 | |
XM_007635562.1 | XP_007633752.1 | 5 - 6 | 1032 | 78 | ||
XM_007635566.1 | XP_007633756.1 | 6 - 7 | 1256 | 78 | ||
XM_007635575.1 | XP_007633765.1 | 5 - 6 | 1172 | 78 | ||
GenBank mRNA | JI879567.1 | - | 1 - 2 | 209 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Probe spans exons |