Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Angptl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003496150.2 | XP_003496198.1 | 5 - 6 | 703 | 96 | |
XM_007647313.1 | XP_007645503.1 | 3 - 4 | 508 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Probe spans exons |