Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Frmpd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007647825.1 | XP_007646015.1 | 4 - 5 | 472 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Probe spans exons |